Skip to Content
Merck
All Photos(1)

Key Documents

T40207

Sigma-Aldrich

o-Tolylhydrazine hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NHNH2·HCl·xH2O
CAS Number:
Molecular Weight:
158.63 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103

mp

187-190 °C (dec.) (lit.)

SMILES string

O.Cl.Cc1ccccc1NN

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service