Skip to Content
Merck
All Photos(1)

Key Documents

T32654

Sigma-Aldrich

2-Thiophenemethanol

98%

Synonym(s):

2-(Hydroxymethyl)thiophene, 2-Thienyl carbinol, Thenyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6OS
CAS Number:
Molecular Weight:
114.17
Beilstein:
383595
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.564 (lit.)

bp

207 °C (lit.)

density

1.205 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCc1cccs1

InChI

1S/C5H6OS/c6-4-5-2-1-3-7-5/h1-3,6H,4H2

InChI key

ZPHGMBGIFODUMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nobuo Ochiai et al.
Journal of chromatography. A, 1628, 461475-461475 (2020-08-22)
For successful profiling of aroma carriers in food samples, a highly efficient extraction method is mandatory. A two-step stir bar sorptive extraction (SBSE) approach, namely fractionated SBSE (Fr-SBSE), was developed to improve both the organoleptic and the chemical identification of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service