Skip to Content
Merck
All Photos(1)

Key Documents

T25003

Sigma-Aldrich

Tetranitromethane

Synonym(s):

NSC 16146, TNM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(NO2)4
CAS Number:
Molecular Weight:
196.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

8.4 mmHg ( 20 °C)

refractive index

n20/D 1.438 (lit.)

bp

126 °C (lit.)

mp

13-14 °C (lit.)

density

1.637 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)C([N+]([O-])=O)([N+]([O-])=O)[N+]([O-])=O

InChI

1S/CN4O8/c6-2(7)1(3(8)9,4(10)11)5(12)13

InChI key

NYTOUQBROMCLBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Nitration of electrophilic alkenes
  • Reactions with B-alkylcatecholboranes forming oxime ethers
  • Heterocyclization of electrophilic alkenes
  • Ring opening of aziridines
  • Acts as an electron acceptor in the mechanism of aerobic visible light formic acid oxidation
  • Reactions with vinyl ketones
Reagent for nitration of enol silyl ethers[1] and aromatic compounds.[2] Employed in the photooxidation of sulfides to sulfoxides.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Ox. Liq. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vaibhav Gupta et al.
The Journal of thoracic and cardiovascular surgery, 156(2), 847-856 (2018-07-18)
Clinical, pathological, and molecular information combined with cancer stage in prognostication algorithms can offer more personalized estimates of survival, which might guide treatment choices. Our aim was to evaluate the quality of prognostication tools in esophageal cancer. We systematically searched
Valerie W Rusch et al.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer, 11(12), 2112-2119 (2016-10-01)
The M component and TNM stage groupings for malignant pleural mesothelioma (MPM) have been empirical. The International Association for the Study of Lung Cancer developed a multinational database to propose evidence-based revisions for the eighth edition of the TNM classification
Synthesis, 1057-1057 (1993)
Tetrahedron Letters, 34, 1859-1859 (1993)
Rui Zhou et al.
EBioMedicine, 42, 420-430 (2019-03-29)
Tumour microenvironment (TME) is critical for the regulation of cancer development as well as therapy. The objective of the current study was the development of a robust prognostic model based on TME-relevant genes. Five public microarray datasets providing clinical information

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service