Skip to Content
Merck
All Photos(1)

Key Documents

T1531

Sigma-Aldrich

Tetrabutylammonium phosphate monobasic

reagent grade, ~98%

Synonym(s):

Tetrabutylammonium dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N[OP(OH)2O]
CAS Number:
Molecular Weight:
339.45
Beilstein:
5196532
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

reagent grade

Assay

~98%

mp

151-154 °C (lit.)

solubility

H2O: soluble, clear to hazy

storage temp.

2-8°C

SMILES string

OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H3O4P/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H3,1,2,3,4)/q+1;/p-1

InChI key

ARRNBPCNZJXHRJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Ion-associating reagent for HPLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K G Wahlung
Journal of chromatography, 115(2), 411-422 (1975-12-24)
A reversed-phase partition chromatographic system for separation of organic anions as ion pairs with quaternary ammonium ions has been developed. Commercial, hydrophobized silica supports are used with 1-pentanol as stationary phase and aqueous solutions of tetrabutylammonium (TBA) as mobile phase.
Pieter van Dillewijn et al.
Applied and environmental microbiology, 74(21), 6820-6823 (2008-09-16)
Homogenous preparations of XenB of Pseudomonas putida, pentaerythritol tetranitrate reductase of Enterobacter cloacae, and N-ethylmaleimide reductase of Escherichia coli, all type II hydride transferases of the Old Yellow Enzyme family of flavoproteins, are shown to reduce the polynitroaromatic compound 2,4,6-trinitrotoluene
Chao Zhong et al.
Carbohydrate polymers, 94(1), 38-45 (2013-04-03)
In this article, a novel and high efficient solvent, tetra-n-Butylammonium Hydroxide (TBAH), was used for dissolution and isolation of straw cellulose from wheat straw. The composition analysis with gas chromatography (GC) and the spectroscopic characterization analysis conducted by X-Ray diffraction
Zhenfeng He et al.
Dalton transactions (Cambridge, England : 2003), 41(33), 10043-10051 (2012-05-29)
A new kind of organic-inorganic hybrid complexes based on polyoxometalate were synthesized through symmetrically grafting two adeninyl groups onto Anderson-type MnMo(6) clusters and encapsulating the clusters by organic surfactants. The resultant complexes exhibited thermal-induced dynamic self-assembly behaviors which greatly depended
David J Posson et al.
Nature structural & molecular biology, 20(2), 159-166 (2012-12-25)
Understanding how ion channels open and close their pores is crucial for comprehending their physiological roles. We used intracellular quaternary ammonium blockers, electrophysiology and X-ray crystallography to locate the voltage-dependent gate in MthK potassium channels from Methanobacterium thermoautotrophicum. Blockers bind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service