Skip to Content
Merck
All Photos(1)

Key Documents

SYX00229

Sigma-Aldrich

9-Methyl-9H-fluorene

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$230.00

$230.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 G
$230.00

About This Item

Empirical Formula (Hill Notation):
C14H12
CAS Number:
Molecular Weight:
180.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

$230.00

PRICE WITHOUT NATIONAL TAXES

Ships within 2 business days(Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

solid

SMILES string

CC1c2ccccc2-c3ccccc13

InChI

1S/C14H12/c1-10-11-6-2-4-8-13(11)14-9-5-3-7-12(10)14/h2-10H,1H3

InChI key

ZVEJRZRAUYJYCO-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of Synthonix

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Chen et al.
Archives of biochemistry and biophysics, 238(1), 195-205 (1985-04-01)
9-Methylfluorene was metabolized by rat liver microsomes to 9-hydroperoxy-9-methylfluorene and 9-hydroxy-9-methylfluorene. The results were confirmed by using a reconstituted cytochrome P-450 oxygenase system purified from phenobarbital-induced rat liver which established its involvement. SKF-525A strongly inhibited the formation of both oxygenation
C Chen et al.
Archives of biochemistry and biophysics, 238(1), 187-194 (1985-04-01)
9-Methylfluorene was found to be oxygenated to 9-hydroperoxy-9-methylfluorene and 9-hydroxy-9-methylfluorene by cytochrome P-450 in the presence of cumene hydroperoxide. Molecular oxygen is required and carbon monoxide is inhibitory. The reaction is inhibited by SKF-525A and metyrapone. Metyrapone and cumene hydroperoxide
E J Lavoie et al.
Mutation research, 91(3), 167-176 (1981-05-01)
Methylated fluorenes were assayed for mutagenic activity towards Salmonella typhimurium TA98 and TA100. None of these methylfluorenes were mutagenic in the absence of metabolic activation. In the presence of 9000 X g supernatant from Aroclor-induced rats, 9-methylfluorene and 1,9-dimethylfluorene were
J E Rice et al.
Journal of toxicology and environmental health, 21(4), 525-532 (1987-01-01)
The genotoxicity of 1-methylpyrene, 1,6-dimethylpyrene, 1-methylfluorene, 9-methylfluorene, and 1,9-dimethylfluorene was evaluated in the hepatocyte primary culture/DNA repair test, and the tumorigenic potency of these compounds was tested by bioassay in newborn mice. With the exception of 1-methylfluorene, all of these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service