Key Documents
SYNS531018
Synthware PTFE stopper with extraction nut
joint: ST/NS 10/18
Synonym(s):
Synthware S531018
Select a Size
1 EA
$60.90
$60.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$60.90
About This Item
UNSPSC Code:
41122500
NACRES:
NB.51
$60.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS10/18
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S531018
General description
PTFE stopper has a Nylon extraction nut permitting easy removal of the stopper from the glass outer joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)