Key Documents
SYNS521000
Synthware storage flask with #15 o-ring joint and high vacuum valve
capacity 1,000 mL
Synonym(s):
Synthware S521000
Select a Size
1 EA
$164.00
$164.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$164.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$164.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass flask
round bottom
single-neck
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S521000
capacity
1,000 mL
General description
Flask has a #15 O-ring joint on the sidearm and a high vacuum valve to control gas or liquid flow into the vessel. Supplied complete with one #116 Viton® O-ring.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)