Documents
SYNS342565
Synthware high vacuum oblique glass stopcock
bore size 8 mm, plug size 25/65
Synonym(s):
Synthware S342565
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S342565
bore size
8 mm
General description
Precision ground with corresponding numbers on the barrel and plug. Lower vacuum chamber is evacuated via a separate 2 mm hole in the plug which is not in the line of flow.
Legal Information
Synthware is a trademark of Kemtech America Inc.