Key Documents
SYNS211944
Synthware double oblique PTFE stopcock plug
bore size 2 mm, Plug Size: 12.9/44
Synonym(s):
Synthware S211944, t-shape stopcock plug
Select a Size
1 EA
$38.60
$38.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$38.60
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$38.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S211944
bore size
2 mm
Related Categories
General description
Synthware double oblique bore stopcock has diagonal, parallel bores that allow the connection of a single stem to both stems on the opposite side.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)