Key Documents
SYNS181635
Synthware straight bore PTFE stopcock plug
bore size 6 mm, Plug Size: 16/35
Synonym(s):
Synthware S181635
Select a Size
1 EA
$43.10
$43.10
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$43.10
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$43.10
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S181635
bore size
6 mm
Related Categories
General description
This replacement kit is complete with one PTFE plug (1:5 taper), one PTFE washer, one O-ring.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)