Key Documents
SYNM260004
Synthware all-glass vacuum/inert gas manifold with solid high vacuum stopcocks
number of ports, 4, Hose Connections: Front-left-right, Rear-left
Synonym(s):
Synthware M260004, vacuum manifold, vacuum unit
Select a Size
1 EA
$948.00
$948.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$948.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
$948.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
number of ports 4
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M260004
General description
With 4 mm double oblique, high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Solid glass stopcocks have constant pressure retention devices. Beams are 22 mm O.D. with front-left-right and rear-left 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)