Key Documents
SYNM101003
Synthware Vacuum/Inert Gas Manifold with double bank and high vacuum valves
number of ports, 3, Hose Connections: Front-left, Rear-right
Synonym(s):
Synthware M101003, vacuum manifold, vacuum unit
Select a Size
1 EA
$721.00
$721.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$721.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
$721.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
number of ports 3
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M101003
General description
General purpose double bank manifold for use with vacuum and inert gas. Supplied with 0-4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and have all banks open. Ports are spaced 100 mm center to center and have an O.D. of 10 mm. With front-left and rear-right 10 mm hose connections.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)