Key Documents
SYNF68272L
Synthware Sure-Grip separatory funnel
2000 mL, stopper size #27
Synonym(s):
Synthware F68272L
Select a Size
1 EA
$317.00
$317.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$317.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
$317.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F68272L
capacity
2000 mL
2000 mL
General description
Globe shaped separatory funnel with a long stopper neck and straight section between the funnel body and stopcock, which provides a secure grip while shaking and also aids in clamping. Available with PTFE stopcocks. Supplied complete with one listed size PTFE or glass stopper.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)