Key Documents
SYNF611425
Synthware pressure equalizing addition funnel with metering stopcock
25 mL, joint: ST/NS 14/20
Synonym(s):
Synthware F611425, dropping funnel
Select a Size
1 EA
$493.00
$493.00
Estimated to ship onMay 24, 2025
Select a Size
Change View
1 EA
$493.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
$493.00
Estimated to ship onMay 24, 2025
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F611425
capacity
25 mL
25 mL
General description
With PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from full flow to drop wise. The metering plug completely solves the problems of sudden overflow or non-flow during addition of reagents.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)