SYNF549100
Synthware™ reaction flask with high vacuum valve
100 mL, joint: ST/NS 19/22, valve size 0-4 mm
Synonym(s):
Synthware F549100, Schlenk flask
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass flask
round bottom
single-neck (high vaccum valve)
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F549100
capacity
100 mL
100 mL
valve size
0-4 mm
Related Categories
General description
Supplied with high vacuum valve with PTFE O-ring at tip and PTFE wiper to protect Viton® O-ring on shaft.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company