Key Documents
SYNF411925
Synthware two neck round bottom flask with angled side neck
25 mL, center joint: ST/NS 19/22, side joint: ST/NS 14/20
Synonym(s):
Synthware F411925, 2-neck round flask
Select a Size
1 EA
$125.00
$125.00
Estimated to ship onApril 26, 2025
Select a Size
Change View
1 EA
$125.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$125.00
Estimated to ship onApril 26, 2025
material
borosilicate glass flask
round bottom
two-neck (angled side neck)
Joint
center joint: ST/NS19/22
side joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F411925
capacity
25 mL
25 mL
General description
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)