Key Documents
SYNF321025
Synthware pear shaped flask
25 mL, joint: ST/NS 14/10
Synonym(s):
Synthware F321025, pear shape glass flask
Select a Size
1 EA
$30.80
$30.80
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$30.80
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$30.80
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS14/10
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F321025
capacity
25 mL
25 mL
General description
Supplied with one compression cap, one Viton® O-ring and one PTFE –faced silicone rubber septum. For replacement caps, see RC112; for Viton® O-rings, see R014; for septa, see R1420.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)