Key Documents
SYNF314200
Synthware recovery flask
200 mL, joint: ST/NS 24/40
Synonym(s):
Synthware F314200, oval shape flask
Select a Size
1 EA
$87.10
$87.10
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$87.10
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$87.10
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F314200
capacity
200 mL
200 mL
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)