Key Documents
SYNF301250
Synthware single-neck round-bottom flask
250 mL, joint: ST/NS 14/20
Synonym(s):
Synthware F301250, glass round flask
Select a Size
1 EA
$67.70
$67.70
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$67.70
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$67.70
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F301250
capacity
250 mL
250 mL
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)