Key Documents
SYNE103424
Synthware Soxhlet extractor body
top joint: ST/NS 34/45, bottom joint: ST/NS 24/40
Synonym(s):
Synthware E103424, glass extractor
Select a Size
1 EA
$183.00
$183.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$183.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
$183.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
bottom joint: ST/NS24/40
joint: ST/NS top34/45
packaging
pkg of 1 ea
manufacturer/tradename
Synthware E103424
Related Categories
General description
Extractor body only.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)