Key Documents
SYND222434
Synthware Abderhalden drying chamber
top joint: ST/NS 24/40, bottom joint: ST/NS 24/40, chamber side joint: ST/NS 34/45
Synonym(s):
Synthware D222434
Select a Size
1 EA
$254.00
$254.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$254.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
$254.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
bottom joint: ST/NS24/40
side joint: ST/NS chamber34/45
top joint: ST/NS24/40
manufacturer/tradename
D222434
Related Categories
General description
Synthware Abderhalden drying chamber is designed for drying samples for elemental analysis. The sample is placed in a vial, which is then placed into the drying chamber. It operates under vacuum with gentle heat created by refluxing low boiling point solvents in the outer jacket.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)