Key Documents
SYNC574200
Synthware water jacketed distillation column with removable hose connections
joint: ST/NS 24/40, jacket H 200 mm
Synonym(s):
Synthware C574200, glass distilling column
Select a Size
1 EA
$214.00
$214.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 EA
$214.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$214.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C574200
jacket H
200 mm
Related Categories
General description
Indentations at the bottom for supporting packing material. Jacket permits the column to be used as a condenser.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)