Key Documents
SYNC564225
Synthware coil-style reflux condenser with removable hose connections
Synthware™ coil-style reflux condenser with removable hose connections
joint: ST/NS 24/40, hose size 10 mm
Synonym(s):
Synthware C564225, coil-type condenser
Select a Size
1 EA
$261.00
$261.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$261.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$261.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C564225
hose size
10 mm
Related Categories
General description
Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Active Filters
  1. 1 Ratings-Only Review