SYNC382630
Synthware™ chromatography column with reservoir, spherical joint and PTFE stopcock
100 mL, joint: ST/NS 35/20, I.D. × L 20.0 mm × 305 mm
Synonym(s):
Synthware C382630, glass chromatograpy column
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS35/20
grade
for chromatography
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C382630
technique(s)
HPLC: suitable
I.D. × L
20.0 mm × 305 mm
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
With 35/20 spherical joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service