Key Documents
SYNC151200
Synthware West condenser
joint: ST/NS 14/20, jacket L 200 mm
Synonym(s):
Synthware C151200, glass West condenser
Select a Size
1 EA
$184.00
$184.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$184.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$184.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C151200
jacket L
200 mm
Related Categories
General description
With narrow annular space for fast coolant speed, thus high cooling efficiency.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)