Key Documents
SYNA961922
Synthware distillation drip adapter
joint: ST/NS 19/22, bend: 105 deg
Synonym(s):
Synthware 961922, glass vacuum adapter
Select a Size
1 EA
$70.30
$70.30
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$70.30
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$70.30
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A961922
Related Categories
General description
With an outer joint at top. Tapered drip tube extends into open receivers.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)