Key Documents
SYNA921922
Synthware vacuum or inert gas adapter
male joint: ST/NS 19/22
Synonym(s):
Synthware A921922, glass vacuum adapter
Select a Size
1 EA
$52.00
$52.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$52.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$52.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
male joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A921922
Related Categories
General description
Synthware vacuum or gas inlet adapter. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection (8 mm O.D.) is at a 90 degree angle to the standard taper inner joint (19/22).
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)