Key Documents
SYNA892440
Synthware distilling adapter with 105 degree bend
top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Synonym(s):
Synthware 892440, bent distillation adapter
Select a Size
1 EA
$62.60
$62.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$62.60
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$62.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
bottom joint: ST/NS24/40
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A892440
Related Categories
General description
With an inner drip joint at the bottom and an outer joint at the top.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)