Key Documents
SYNA872440
Synthware 75 degree angled drying tube
joint: ST/NS 24/40
Synonym(s):
Synthware A872440, glass drying tube, Drying tube 75° bent
Select a Size
1 EA
$56.60
$56.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$56.60
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$56.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
colorless tube
glass tube
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A872440
General description
Angled at 75° to avoid desiccant contact with reactions.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)