Key Documents
SYNA831420
Synthware inlet adapter with fritted disc
joint: ST/NS 14/20
Synonym(s):
Synthware A831420, glass inlet adapter
Select a Size
1 EA
$86.50
$86.50
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$86.50
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$86.50
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A831420
General description
With an extra coarse disc to prevent solid from being introduced into reaction system during liquid transfer.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)