Key Documents
SYNA342440
Synthware twin hose connecting adapter
joint: ST/NS 24/40
Synonym(s):
Synthware A342440, Adapter, twin connecting hose
Select a Size
1 EA
$32.20
$32.20
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$32.20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$32.20
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A342440
Related Categories
General description
Adapter has two hose connections 180° from one another to permit a flow of inert gas over the apparatus.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)