Key Documents
SYNA302440
Synthware vacuum or argon/nitrogen adapter with hose connection on top
joint: ST/NS 24/40
Synonym(s):
Synthware A302440, inert gas adapter, vacuum adapter
Select a Size
1 EA
$51.30
$51.30
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$51.30
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$51.30
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A302440
Related Categories
General description
With hose connection on the top and male joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)