Key Documents
SYNA253424
Synthware enlarging adapter
top joint: ST/NS 34/45, bottom joint: ST/NS 24/40
Synonym(s):
Synthware A253424, glass connecting adapter
Select a Size
1 EA
$50.50
$50.50
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Select a Size
Change View
1 EA
$50.50
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$50.50
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Joint
bottom joint: ST/NS24/40
joint: ST/NS top34/45
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A253424
Related Categories
General description
Synthware enlarging adapter is used for connecting dissimilar standard taper joint sizes. The top joint (34/45) is larger that the bottom joint (24/40).
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)