Key Documents
SYNA232424
Synthware anti-splash adapter with fritted disc
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Synonym(s):
Synthware A232424, rotary evaporator bump trap
Select a Size
1 EA
$172.00
$172.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$172.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$172.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
bottom joint: ST/NS24/40
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A232424
capacity
100 mL
Related Categories
General description
With extra coarse fritted disc sealed above lower inner joint. Frit reduces foam from being carryover.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)