Key Documents
SYNA202434
Synthware reducing adapter
top joint: ST/NS 24/40, bottom joint: ST/NS 34/45
Synonym(s):
Synthware A202434, glass reducing adapter, connecting adapter
Select a Size
1 EA
$47.30
$47.30
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$47.30
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$47.30
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
bottom joint: ST/NS34/45
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A202434
Related Categories
General description
Synthware reducing adapter is used for connecting dissimilar standard taper joint sizes. The bottom joint (34/45) is larger than the top joint (24/40).
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)