Skip to Content
Merck
All Photos(1)

Key Documents

SM40100201

Sigma-Aldrich

SmartPor, alumina membrane

thickness 100 μm, pore diameter 40 nm, pore size 125 nm (interpore distance), size 20 mm × 20 mm, pkg of 1 pieces

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$322.00

$322.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$322.00

About This Item

Linear Formula:
Al
UNSPSC Code:
12352103
NACRES:
NA.23

$322.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

material

alumina membrane

description

Pore geometry: trigonal arrangement

packaging

pkg of 1 pieces

size

20 mm × 20 mm

thickness

100 μm

pore size

125 nm (interpore distance)
40 nm pore diameter

compatibility

(Pore geometry: trigonal arrangement)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service