Skip to Content
Merck
All Photos(1)

Key Documents

SLW4546/45

Pyrex® Spare sealing disc for high temperature screwcaps

natural, GL 45

Synonym(s):

Pyrex, SciLabware 4546/45

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

material

PTFE face
natural

manufacturer/tradename

4546/45
SciLabware

fitting

GL 45

Looking for similar products? Visit Product Comparison Guide

General description

For use with SLW4526 screwcaps.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service