SLW1515/16D
Pyrex® Reagent bottles, round bottom with reusable screw caps
capacity 5000 mL
Synonym(s):
Pyrex, SciLabware 1515/16D
Select a Size
$390.00
Select a Size
About This Item
$390.00
Recommended Products
material
PTFE seal
Pyrex bottle
black PBT cap
colorless bottle
round bottle
round bottom
white graduation
Joint
Joints threaded neck (SVL 60)
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
SciLabware 1515/16D
bottle diam. × H
181 mm × 324 mm
cap size
60 mm
capacity
5000 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Complies with ISO 4796-1
- Ideal for storage of reagents, analytical standards and culture media etc.
- Manufactured from Pyrex borosilicate glass
- Excellent resistance to chemical attack and high temperatures
- Supplied with PBT screwcaps having a PTFE faced sealing disc
- Withstands hot air sterilisation up to 180 deg. C
- Autoclavable
- White graduations and marking spot
Features and Benefits
- Manufactured from PYREX® borosilicate glass
- Complies with ISO 4796-1
- Ideal for storage of reagents, analytical standards and culture media etc.
- Excellent resistance to chemical attack and high temperatures
- Supplied with PBT screw caps having a PTFE faced cap liner
- Withstands hot air sterilisation up to 180 °C
- Autoclavable
- White graduations and marking spot
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service