Skip to Content
Merck
All Photos(1)

Key Documents

QBD11626

Sigma-Aldrich

MAL-dPEG®4-Glu(TFP ester)-NH-dPEG®4-Glu(TFP ester)-NH-m-dPEG®24

>90% (HPLC)

Synonym(s):

MAL-PEG4-Glu(TFP ester)-NH-PEG4-Glu(TFP ester)-NH-m-PEG24, Sidewinder TFP-PEG-maleimide

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$1,310.00

$1,310.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$1,310.00

About This Item

Empirical Formula (Hill Notation):
C100H162F8N6O43
Molecular Weight:
2288.36
UNSPSC Code:
12352108
NACRES:
NA.22

$1,310.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>90% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: Y-shape
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

MAL-dPEG4-Glu(TFP ester)-NH-dPEG4-Glu(TFP ester)-NH-m-dPEG24 is a novel, patent-pending Sidewinder product from Quanta BioDesign that permits attachment of hydrophobic groups on the sidearms of two glutamic acid moieties. Short hydrophilic spacers separate the glutamates, and a long hydrophilic dPEG unit terminates the construct. A sulfhydryl-reactive maleimide group provides an attachment point for the construct to another molecule, such as a protein or peptide. Tetrafluorophenyl esters functionalize the two glutamate units along the Sidewinder backbone.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
Sidewinder is a trademark of Quanta BioDesign
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service