Skip to Content
Merck
All Photos(1)

Key Documents

QBD11414

Sigma-Aldrich

Tetra(-amido-dPEG ®23-MAL)pentaerythritol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$2,180.00
100 MG
$3,640.00

$2,180.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$2,180.00
100 MG
$3,640.00

About This Item

Empirical Formula (Hill Notation):
C237H440N12O112
Molecular Weight:
5250.04
UNSPSC Code:
51171641
NACRES:
NA.22

$2,180.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reactivity: thiol reactive

polymer architecture

shape: branched
functionality: homofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

Tetra(-amido-dPEG®₂₃-MAL)pentaerythritol has four connected dPEG spacers that are each capped with a maleimide group which can react with a free thiol.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service