Skip to Content
Merck
All Photos(1)

Key Documents

QBD11389

Sigma-Aldrich

t-Boc-N-amido-dPEG®24-amido-dPEG®23-amine

>95% (HPLC)

Synonym(s):

t-Boc-N-amido-PEG24-amido-PEG23-amine, t-Boc-PEG-amine, t-Boc-PEG47-NH2

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$921.00

$921.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$921.00

About This Item

Empirical Formula (Hill Notation):
C104H209N3O50
Molecular Weight:
2301.76
UNSPSC Code:
12352116
NACRES:
NA.22

$921.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: homobifunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

t-Boc-dPEG24-amido-dPEG23-amine is an increasingly popular choice for homobifunctional crosslinking. A tert-butyl group protects one end of this diamine-functionalized monodispersed PEG crosslinker. Carboxylic acids, aldehydes, and ketones react with both ends of the crosslinker. Carboxylic acids form amide bonds, while aldehydes and ketones react to form Schiff bases that are reducible to secondary amines. The mono-protected end of the molecule gives the user tighter control over crosslinking. The single molecular weight, discrete chain-length PEG linker is 149 atoms (107.7 Å) long and is the monodisperse PEG equivalent to PEG2000.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service