Skip to Content
Merck
All Photos(1)

Documents

QBD11094

Sigma-Aldrich

m-dPEG®12-amido-dPEG®24-DSPE

>95% (HPLC)

Synonym(s):

m-PEG12-amido-PEG24-DSPE, DSPE-PEG36-OMe, mPEG-DSPE, mPEG1500-DSPE, mPEG36-DSPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C118H233N2O46P
Molecular Weight:
2447.07
UNSPSC Code:
12352200
NACRES:
NA.22

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

m-dPEG12-amido-dPEG24-DSPE is designed for use in liposomes and micelles. The product contains a lipophilic tail of 1, 2-Distearoyl-sn-glycero-3-phosphoethanolamine (DSPE) and a hydrophilic head consisting of a methoxy-terminated, single molecular weight, discrete polyethylene glycol (dPEG). The dPEG portion of the molecule is 117 atoms (76.364 Å) long with a molecular weight of 1744 Daltons. The molecular weight of the full molecule is 2491 Daltons. m-dPEG12-amido-dPEG24-DSPE is a monodisperse equivalent of the polymeric PEG1500.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service