Skip to Content
Merck
All Photos(1)

Key Documents

QBD11007

Sigma-Aldrich

Fmoc-N-amido-dPEG ®24-TFP ester

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$674.00
1000 MG
$2,640.00

$674.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$674.00
1000 MG
$2,640.00

About This Item

Empirical Formula (Hill Notation):
C72H113F4NO28
Molecular Weight:
1516.65
UNSPSC Code:
51171641
NACRES:
NA.22

$674.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations
reagent type: cross-linking reagent

functional group

Fmoc

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(OCC1C(C=CC=C2)=C2C3=C1C=CC=C3)=O)OC4=C(F)C(F)=CC(F)=C4F

InChI key

PFGUQFPJXCYGKT-UHFFFAOYSA-N

Features and Benefits

Fmoc-N-amido-dPEG®24-TFP ester can be used to introduce a dPEG linker and TFP ester during peptide synthesis.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service