Skip to Content
Merck
All Photos(1)

Key Documents

QBD10822

Sigma-Aldrich

Biotin-dPEG®11-Lipoamide

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$649.00

$649.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$649.00

About This Item

Empirical Formula (Hill Notation):
C42H78N4O14S3
CAS Number:
Molecular Weight:
959.28
MDL number:
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

$649.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: Biotinylations
reactivity: gold reactive

reagent type: cross-linking reagent

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCCCC1SSCC1)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(CCCCC2SCC(C2N3)NC3=O)=O

InChI

1S/C42H78N4O14S3/c47-39(7-3-1-5-36-9-34-62-63-36)43-10-12-50-14-16-52-18-20-54-22-24-56-26-28-58-30-32-60-33-31-59-29-27-57-25-23-55-21-19-53-17-15-51-13-11-44-40(48)8-4-2-6-38-41-37(35-61-38)45-42(49)46-41/h36-38,41H,1-35H2,(H,43,47)(H,44,48)(H2,45,46,49

InChI key

LHDRNWLHXCGFBV-UHFFFAOYSA-N

Features and Benefits

Biotin-dPEG®11-lipoamide is designed to provide biotinylation of metallic (e.g., gold) surfaces for use in diverse life science and nanotechnology applications. The single molecular weight dPEG® spacer is 52.9 Å long. It is hydrophilic and non-immunogenic. The lipoic acid group forms two strong, stable dative bonds to metal surfaces per molecule.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service