Skip to Content
Merck
All Photos(1)

Documents

QBD10492

Sigma-Aldrich

Aminooxy-dPEG®12-amido-dPEG®12-(m-dPEG®11)3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C136H270N6O66
Molecular Weight:
3045.60
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chemical modification reagent
reagent type: cross-linking reagent
reactivity: carbonyl reactive

polymer architecture

shape: branched
functionality: homofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(NC(COCCC(NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)(COCCC(NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)COCCC(NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(CCOCCON)=O

Features and Benefits

Aminooxy-dPEG®12-amido-dPEG®12-(m-dPEG®11)3 is a branched dPEG® reagent that is highly useful for protein and peptide modification to increase hydrodynamic volume and serum half-life and to greatly reduce the immunogenicity of the modified peptide or protein. This dPEG® product consists of a linear "tail" with a reactive group, a tris core to provide branching, and methoxy-terminated dPEG® branches. The numbers following dPEG® are the exact number of repeat units in the linear chain. This dPEG® product is hydrophilic, non-immunogenic, single molecular weight compound. The aminooxy functional group reacts with aldehydes and ketones to form oxime bonds.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service