Skip to Content
Merck
All Photos(1)

Key Documents

QBD10397

Sigma-Aldrich

Bis-MAL-dPEG®11

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$493.00
1000 MG
$2,690.00

$493.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$493.00
1000 MG
$2,690.00

About This Item

Empirical Formula (Hill Notation):
C38H62N4O17
CAS Number:
Molecular Weight:
846.92
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.22

$493.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reactivity: thiol reactive

polymer architecture

shape: linear
functionality: homobifunctional

shipped in

ambient

storage temp.

−20°C

Application

Bis-MAL-dPEG®11 is useful for producing dimeric molecules after reaction with free thiol groups.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2dPEG is a registered trademark of Quanta BioDesign
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service