Skip to Content
Merck
All Photos(1)

Key Documents

QBD10007

Sigma-Aldrich

Biotin-dPEG®24-TFP ester

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$471.00
1000 MG
$2,650.00

$471.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$471.00
1000 MG
$2,650.00

About This Item

Empirical Formula (Hill Notation):
C67H117F4N3O28S
Molecular Weight:
1520.71
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$471.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: Biotinylations
reaction type: Biotinylations
reactivity: amine reactive

reagent type: cross-linking reagent

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCCCC1SCC(N2)C1NC2=O)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(OC3=C(C(F)=CC(F)=C3F)F)=O

InChI key

DZXHDWHQLGLRLF-UHFFFAOYSA-N

Features and Benefits

The Biotin-dPEG®24-TFP ester has 2,3,5,6-tetrafluorophenyl (TFP) moiety which hydrolyze more slowly than NHS esters in aqueous media and are more reactive towards amines than NHS esters. The single molecular weight Biotin-dPEG®24-TFP ester has a long spacer of precise length (99.3 Å) between the biotin moiety and the labeled molecule. This dPEG spacer imparts a high degree of water solubility to the biotin label. For biotinylated peptides and proteins, the dPEG spacer dramatically reduces aggregation and precipitation of the modified molecule and reduces non-specific binding of the molecule, both of which are common problems with molecules that have been biotinylated with the widely used NHS-LC-biotin.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service