Skip to Content
Merck
All Photos(4)

Key Documents

P8533

Sigma-Aldrich

Tributylammonium pyrophosphate

Synonym(s):

N,N-dibutyl-1-butanamine diphosphate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$84.10
1 G
$235.00
5 G
$757.00

$84.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$84.10
1 G
$235.00
5 G
$757.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$84.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

form

powder

storage temp.

−20°C

SMILES string

OP(O)(=O)OP(O)(O)=O.CCCCN(CCCC)CCCC.CCCCN(CCCC)CCCC

InChI

1S/2C12H27N.H4O7P2/c2*1-4-7-10-13(11-8-5-2)12-9-6-3;1-8(2,3)7-9(4,5)6/h2*4-12H2,1-3H3;(H2,1,2,3)(H2,4,5,6)

InChI key

WRMXOVHLRUVREB-UHFFFAOYSA-N

General description

Tributylammonium pyrophosphate is a phosphorylating reagent used in the synthesis of modified dNTPs (deoxynucleoside triphosphates) which are important building blocks for the generation of DNA, RNA, functionalized nucleic acids.[1]

Application

Reactant for:
  • Enzymic synthesis of 3′-deoxy-apio-nucleic acid duplexes
  • Synthesis of deoxynucleoside S-methylphosphonic acids
  • Preparation of fluorescent non-nucleotide ATP analog N-methylanthraniloylamideethyl triphosphate
Tributylammonium pyrophosphate used as a phosphorylating reagent:
  • In the synthesis of S′-deoxy-S′-methylphosphonic acid diphosphate along with 1, 1′-carbonyl diimidazole.
  • In the synthesis of gram-scale level base-modified ribonucleoside-5′-O-triphosphates.
  • In one-pot chemical synthesis of 2′-deoxynucleoside-5′-triphosphates with better protection.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A facile and effective synthesis of dinucleotide 5?-triphosphates
Abramova T, et al.
Bioorganic & Medicinal Chemistry, 15, 6549-6555 (2007)
CONVERSION OF MONO- AND OLIGODEOXYRIBONUCLEOTIDES TO 5-TRIPHOSPHATES.
D E HOARD et al.
Journal of the American Chemical Society, 87, 1785-1788 (1965-04-20)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service