Skip to Content
Merck
All Photos(1)

Key Documents

P51

Sigma-Aldrich

Palmitic acid

90%

Synonym(s):

1-Pentadecanecarboxylic acid, C16:0, Cetylic acid, Hexadecanoic acid, NSC 5030, PamOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COOH
CAS Number:
Molecular Weight:
256.42
Beilstein:
607489
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

10 mmHg ( 210 °C)

Assay

90%

bp

271.5 °C/100 mmHg (lit.)

mp

61-62.5 °C (lit.)

density

0.852 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCC(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)

InChI key

IPCSVZSSVZVIGE-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonas Sieber et al.
The American journal of pathology, 183(3), 735-744 (2013-07-23)
Type 2 diabetes mellitus is characterized by dyslipidemia with elevated free fatty acids (FFAs). Loss of podocytes is a hallmark of diabetic nephropathy, and podocytes are highly susceptible to saturated FFAs but not to protective, monounsaturated FFAs. We report that
B Vessby et al.
Diabetologia, 37(10), 1044-1050 (1994-10-01)
Recent data indicate that peripheral insulin sensitivity may be influenced by dietary fat quality and skeletal muscle phospholipid fatty acid composition. During a health survey of 70-year-old men insulin sensitivity was measured by the euglycaemic hyperinsulinaemic clamp technique and the
Hjalti Kristinsson et al.
Endocrinology, 154(11), 4078-4088 (2013-09-17)
Free fatty acids (FFAs) have pleiotropic effects on the pancreatic β-cell. Although acute exposure to FFAs stimulates glucose-stimulated insulin secretion (GSIS), prolonged exposure impairs GSIS and causes apoptosis. FFAs exert their effects both via intracellular metabolism and interaction with the
Sanja Blaskovic et al.
The FEBS journal, 280(12), 2766-2774 (2013-04-05)
S-palmitoylation is post-translational modification, which consists in the addition of a C16 acyl chain to cytosolic cysteines and which is unique amongst lipid modifications in that it is reversible. It can thus, like phosphorylation or ubiquitination, act as a switch.
Siri Malmgren et al.
The Journal of biological chemistry, 288(17), 11973-11987 (2013-03-12)
Lipotoxicity is a presumed pathogenetic process whereby elevated circulating and stored lipids in type 2 diabetes cause pancreatic β-cell failure. To resolve the underlying molecular mechanisms, we exposed clonal INS-1 832/13 β-cells to palmitate for 48 h. We observed elevated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service