Skip to Content
Merck
All Photos(2)

Documents

P23350

Sigma-Aldrich

1,3-Phenylenediacetic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2CO2H)2
CAS Number:
Molecular Weight:
194.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

175-177 °C (lit.)

SMILES string

OC(=O)Cc1cccc(CC(O)=O)c1

InChI

1S/C10H10O4/c11-9(12)5-7-2-1-3-8(4-7)6-10(13)14/h1-4H,5-6H2,(H,11,12)(H,13,14)

InChI key

GDYYIJNDPMFMTB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuseppe Tronci et al.
International journal of biological macromolecules, 81, 112-120 (2015-08-05)
The formation of naturally derived materials with wet stable fibrous architectures is paramount in order to mimic the features of tissues at the molecular and microscopic scale. Here, we investigated the formation of wet-spun fibres based on collagen-derived polypeptides with
Jian-Guo Wang et al.
Bioorganic & medicinal chemistry, 15(1), 374-380 (2006-10-20)
Acetohydroxyacid synthase (AHAS; EC 2.2.1.6) catalyzes the first common step in branched-chain amino acid biosynthesis. This enzyme is an important target for the design of environmental-benign herbicides. Based on the crystal structure of AHAS/sulfonylurea complex, we have carried out computational

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service